Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lysozyme C/LYZ protein, human (HEK293, C-His) | 50UG
SDP

Catalog No. 5000174992
Encompass_Preferred

Recombinant human lysozyme C expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized (50 μg), this ~16 kDa bacteriolytic enzyme hydrolyzes bacterial cell wall polysaccharides and is used for studies of innate immunity, antimicrobial activity, and biochemical or functional assays. Provided with handling and storage guidance for research use only.

  • Recombinant protein expressed in HEK293 cells, preserving mammalian post-translational modifications.
  • C-terminal His tag for straightforward purification and detection.
  • Supplied lyophilized for stable storage and easy reconstitution.
  • Low endotoxin suitable for sensitive biological assays.
  • Documented specific activity to support functional studies.
  • Recommended storage at -20 °C or -80 °C to maintain activity.

Catalog No. 50-001-74992 Supplier Medchemexpress LLC Supplier No. HYP70390A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human lysozyme C expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized (50 μg), this ~16 kDa bacteriolytic enzyme hydrolyzes bacterial cell wall polysaccharides and is used for studies of innate immunity, antimicrobial activity, and biochemical or functional assays. Provided with handling and storage guidance for research use only.

  • Recombinant protein expressed in HEK293 cells, preserving mammalian post-translational modifications.
  • C-terminal His tag for straightforward purification and detection.
  • Supplied lyophilized for stable storage and easy reconstitution.
  • Low endotoxin suitable for sensitive biological assays.
  • Documented specific activity to support functional studies.
  • Recommended storage at -20 °C or -80 °C to maintain activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.