Learn More
Cayman Chemical Lysozyme C MoncLNal ntIbod 1ea
SDP
Catalog No. 502969925
Encompass_Preferred
Host: mouse, clone 4C3 • Isotype: IgG3 • Cross Reactivity: (+) human and mouse lysozyme C • Application(s): FC and IF
Catalog No. 50-296-9925 Supplier Cayman Chemical Supplier No. 156411 EA
Host: mouse, clone 4C3 • Isotype: IgG3 • Cross Reactivity: (+) human and mouse lysozyme C • Application(s): FC and IF