Learn More
Supelco Inc LYSOZYME CHICKEN EGG WHITE 10G
Catalog No. NC9273336
Encompass
Lysozyme, lyophilized powder, protein A%Y90A %, A%Y40,000A units/mg protein
Catalog No. NC9273336 Supplier Supelco Inc Supplier No. L687610G
Lysozyme, lyophilized powder, protein A%Y90A %, A%Y40,000A units/mg protein