Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme, Chicken Egg White, Ultrapure, Thermo Scientific™

Catalog No. AAJ1864522 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1864522 100 g Plastic bottle
AAJ1864509 10 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ1864522 Supplier Thermo Scientific Supplier No. J1864522

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12650-88-3
Molecular Formula C125H196N40O36S2
Molecular Weight (g/mol) 2899.307
MDL Number MFCD00131557
InChI Key JFXJPYIEDZSWNF-JWBGUOTLSA-N
Synonym lysozyme, chicken egg white, unii-1zml031y3c, lysozyme chicken egg white, unii-968jka7t33
PubChem CID 131750181
SMILES CC(C)CC(C(=O)NC(CC(=O)O)C(=O)NC(CC(=O)N)C(=O)NC(CC1=CC=C(C=C1)O)C(=O)NC(CCCNC(=N)N)C(=O)NCC(=O)NC(CC2=CC=C(C=C2)O)C(=O)NC(CO)C(=O)NC(CC(C)C)C(=O)NCC(=O)NC(CC(=O)N)C(=O)O)NC(=O)CNC(=O)C(CC3=CN=CN3)NC(=O)C(CCCNC(=N)N)NC(=O)C(CCCCN)NC(=O)C(CCSC)NC(=O)C(C)NC(

Specifications

Packaging Plastic bottle
Quantity 100 g
Grade Ultrapure
Physical Form Crystalline Powder
Chemical Name or Material Lysozyme
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.