Learn More
Gold Biotechnology Inc Lysozyme Egg White 1 g
SDP
Catalog No. 5000562042
Encompass_Preferred
Lysozyme is an enzyme capable of breaking down the bacterial cell wall and is typically used to enhance protein extractions or nucleic acid extractions It works by hydrolyzing the 1 4- -linkages located between the N- acetylmuramic acid and the N- acetyl-D-glucosamine residues in peptidoglycan
Catalog No. 50-005-62042 Supplier Gold Biotechnology Inc Supplier No. L0401
Lysozyme is an enzyme capable of breaking down the bacterial cell wall and is typically used to enhance protein extractions or nucleic acid extractions It works by hydrolyzing the 1 4- -linkages located between the N- acetylmuramic acid and the N- acetyl-D-glucosamine residues in peptidoglycan