Learn More
Gold Biotechnology Inc Lysozyme, Egg White 5 g
SDP
Catalog No. 501532813
Encompass_Preferred
Lysozyme is an enzyme capable of breaking down the bacterial cell wall, and is typically used to enhance protein extractions or nucleic acid extractions. It works by hydrolyzing the 1,4-β-linkages located between the N-acetylmuramic acid and the N-acetyl-D-glucosamine residues in peptidoglycan.
Catalog No. 50-153-2813 Supplier Gold Biotechnology Inc Supplier No. L0405
Lysozyme is an enzyme capable of breaking down the bacterial cell wall, and is typically used to enhance protein extractions or nucleic acid extractions. It works by hydrolyzing the 1,4-β-linkages located between the N-acetylmuramic acid and the N-acetyl-D-glucosamine residues in peptidoglycan.