Learn More
Gold Biotechnology Inc Lysozyme, Egg White 100 g
SDP
Catalog No. 501532811
Encompass_Preferred
Lysozyme is an enzyme capable of breaking down the bacterial cell wall, and is typically used to enhance protein extractions or nucleic acid extractions. It works by hydrolyzing the 1,4-β-linkages located between the
Catalog No. 50-153-2811 Supplier Gold Biotechnology Inc Supplier No. L040100
Lysozyme is an enzyme capable of breaking down the bacterial cell wall, and is typically used to enhance protein extractions or nucleic acid extractions. It works by hydrolyzing the 1,4-β-linkages located between the