Learn More
Research Products International Corp Lysozyme from Chicken Egg White, Salt Free, 5 Grams
SDP
Catalog No. 50488781
Encompass_Preferred
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.
Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.
Catalog No. 50-488-781 Supplier Research Products International Corp Supplier No. L381005.0
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.
Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.