Learn More
Sigma Aldrich Fine Chemicals Biosciences Lysozyme human recombinant10G
Catalog No. 502902165
Encompass
Lysozyme is present in tears saliva and human milk. The human lysozyme expressed in rice grains is an excellent supplemental source of this antimicrobial protein. Lysozyme is mapped to human chromosome 12q15 and corresponds to a molecular weight of 16.5kDa. It is a positively charged and a C-type lysozyme with and structural domains.
Catalog No. 50-290-2165 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L166710G
Lysozyme is present in tears saliva and human milk. The human lysozyme expressed in rice grains is an excellent supplemental source of this antimicrobial protein. Lysozyme is mapped to human chromosome 12q15 and corresponds to a molecular weight of 16.5kDa. It is a positively charged and a C-type lysozyme with and structural domains.