Learn More
STEMCELL Technologies Anti-Human Lysozyme Antibody, Polyclonal, Size: 100 ul
SDP
Supplier:  STEMCELL Technologies 1001068
Encompass
This rabbit polyclonal antibody reacts with human lysozyme a secreted protein found in high concentration in various tissues and fluids such as in liver articular cartilage saliva and tears. Use this polyclonal antibody in applications requiring lysozyme detection such as immunohistochemistry protocols (IHC-P). It has been verified for use with immunocytochemistry (ICC).
Catalog No. 50-239-0450