Learn More
Carbosynth LLC. Lysyl-Bradykinin (Kallidin) (Human, Bovine) (0.5 mg vial), 1ea, MW: 1188.4, Molecular formula: C56H85N17O12, CAS# 342-10-9
SDP
Supplier:  Carbosynth LLC. PBK4008V0.5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg, Kallidin , Storage Temperature: -20C, References: J.V. Pierce and M.E. Webster, Biochem. Biophys. Res. Commun., 5, 353 (1961). (Original; Human), D.F. Elliott and G.P. Lewis, Biochem. J., 95, 437 (1965). (Seq.; Bovine)
Catalog No. 50-168-6237