Learn More
Medchemexpress LLC Lysyl hydroxylase 2-IN-1 | 3053862-11-3 | 99.9% | 310.35 | 5 MG
SDP
Catalog No. 5000361926
Encompass_Preferred
Lysyl hydroxylase 2-IN-1 is a selective inhibitor of lysyl hydroxylase 2 (LH2), with an IC50 of approximately 300 nM. This compound demonstrates specific selectivity for LH2 over LH1 and LH3, making it a valuable tool for research in related biological pathways.
  • Selective LH2 inhibitor
  • IC50 of approximately 300 nM
  • Demonstrates selectivity over LH1 and LH3
  • Available as a solid, light yellow to yellow in color
Catalog No. 50-003-61926 Supplier Medchemexpress LLC Supplier No. HY1495975MG
Lysyl hydroxylase 2-IN-1 is a selective inhibitor of lysyl hydroxylase 2 (LH2), with an IC50 of approximately 300 nM. This compound demonstrates specific selectivity for LH2 over LH1 and LH3, making it a valuable tool for research in related biological pathways.
  • Selective LH2 inhibitor
  • IC50 of approximately 300 nM
  • Demonstrates selectivity over LH1 and LH3
  • Available as a solid, light yellow to yellow in color