Learn More
Aat Bioquest Amplite™ Fluorimetric Lysyl Oxidase Assay Kit *Red Fluorescence*
SDP
Catalog No. 501953557
Encompass_Preferred
Lysyl oxidase (LOX) is an extracellular enzyme that catalyzes formation of aldehydes from lysine residues in collagen and elastin precursors.
Catalog No. 50-195-3557 Supplier Aat Bioquest Supplier No. 15255
Lysyl oxidase (LOX) is an extracellular enzyme that catalyzes formation of aldehydes from lysine residues in collagen and elastin precursors.