Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lysyl oxidase-like 2 (LOXL2), human recombinant protein | >95.0% | 10 UG
SDP

Catalog No. 5000294489
Encompass_Preferred

Recombinant human lysyl oxidase-like 2 (LOXL2) expressed in HEK293 cells and supplied as a lyophilized C-terminal 6xHis-tagged protein. Intended for biochemical assays and cell biology research; the protein is >95% pure by reducing SDS-PAGE, shows measured enzymatic activity, and is provided in microgram quantities.

  • Expression system: HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS, pH 7.4, or PBS with 8% trehalose.
  • Recommended reconstitution to ≥100 μg/mL; store aliquots at -20°C or -80°C.
  • Measured specific activity >2 pmol/min/μg (H2O2 production assay).
  • Sequence range M26-Q774; approximate molecular weight ~110 kDa.

Catalog No. 50-002-94489 Supplier Medchemexpress LLC Supplier No. HYP7477010UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human lysyl oxidase-like 2 (LOXL2) expressed in HEK293 cells and supplied as a lyophilized C-terminal 6xHis-tagged protein. Intended for biochemical assays and cell biology research; the protein is >95% pure by reducing SDS-PAGE, shows measured enzymatic activity, and is provided in microgram quantities.

  • Expression system: HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation from PBS, pH 7.4, or PBS with 8% trehalose.
  • Recommended reconstitution to ≥100 μg/mL; store aliquots at -20°C or -80°C.
  • Measured specific activity >2 pmol/min/μg (H2O2 production assay).
  • Sequence range M26-Q774; approximate molecular weight ~110 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.