Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lysyl oxidase-like 2 (LOXL2) protein | >95.0% | 50 UG
SDP

Catalog No. 5000294490
Encompass_Preferred

Recombinant human LOXL2 (lysyl oxidase-like 2) produced in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder for research use in extracellular matrix, EMT, and enzymatic activity studies.

  • Expressed in HEK293 cells with a C-terminal 6xHis tag.
  • Glycosylated, full-length mature protein (M26-Q774).
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin less than 1 EU/μg (LAL).
  • Lyophilized from PBS, pH 7.4, optionally with 8% trehalose.
  • Specific activity greater than 2 pmol/min/μg in benzylamine oxidation assay.
  • Recommended storage at -20°C; after reconstitution, 4°C for 1 week or -20°C long term with carrier protein.

Catalog No. 50-002-94490 Supplier Medchemexpress LLC Supplier No. HYP7477050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human LOXL2 (lysyl oxidase-like 2) produced in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder for research use in extracellular matrix, EMT, and enzymatic activity studies.

  • Expressed in HEK293 cells with a C-terminal 6xHis tag.
  • Glycosylated, full-length mature protein (M26-Q774).
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin less than 1 EU/μg (LAL).
  • Lyophilized from PBS, pH 7.4, optionally with 8% trehalose.
  • Specific activity greater than 2 pmol/min/μg in benzylamine oxidation assay.
  • Recommended storage at -20°C; after reconstitution, 4°C for 1 week or -20°C long term with carrier protein.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.