Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lysylglutamic acid | 45234-02-4 | MFCD00038215 | 98.0% | 275.30 g/mol | C11H21N3O5 | 100 MG
SDP

Catalog No. 5000281969
Encompass_Preferred

Lysylglutamic acid is a dipeptide composed of the amino acids lysine and glutamic acid. It is provided as a research-grade endogenous metabolite reagent and has documented interaction with the PEPT1 transporter (Ki = 1.3 mM). The product is supplied as a solid with defined purity and storage conditions for laboratory use.

  • CAS number: 45234-02-4
  • Molecular formula: C11H21N3O5
  • Molecular weight: 275.30 g/mol
  • Purity: 98.0%
  • Appearance: solid
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Reported activity: PEPT1 transporter interaction, Ki = 1.3 mM

Catalog No. 50-002-81969 Supplier Medchemexpress LLC Supplier No. HYW327145100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Lysylglutamic acid is a dipeptide composed of the amino acids lysine and glutamic acid. It is provided as a research-grade endogenous metabolite reagent and has documented interaction with the PEPT1 transporter (Ki = 1.3 mM). The product is supplied as a solid with defined purity and storage conditions for laboratory use.

  • CAS number: 45234-02-4
  • Molecular formula: C11H21N3O5
  • Molecular weight: 275.30 g/mol
  • Purity: 98.0%
  • Appearance: solid
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Reported activity: PEPT1 transporter interaction, Ki = 1.3 mM

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.