Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lysylglutamic acid | 45234-02-4 | MFCD00038215 | 98.0% | 275.30 g/mol | C11H21N3O5 | 50 MG
SDP

Catalog No. 5000280692
Encompass_Preferred

Lysylglutamic acid is a dipeptide composed of the amino acids lysine and glutamic acid supplied for laboratory research. The compound has molecular formula C11H21N3O5, molecular weight 275.30 g/mol, and CAS 45234-02-4. A reported Ki of 1.3 mM for the PEPT1 membrane transporter is documented. It is provided as a high-purity reagent for biochemical and transporter studies.

  • Purity 98.0%.
  • Molecular weight 275.30 g/mol and formula C11H21N3O5.
  • Reported Ki 1.3 mM for PEPT1 transporter.
  • Available in multiple small-scale package sizes for laboratory use.
  • Intended for research use only; not for human consumption.
  • Suitable for peptide research, transporter assays, and biochemical studies.

Catalog No. 50-002-80692 Supplier Medchemexpress LLC Supplier No. HYW32714550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Lysylglutamic acid is a dipeptide composed of the amino acids lysine and glutamic acid supplied for laboratory research. The compound has molecular formula C11H21N3O5, molecular weight 275.30 g/mol, and CAS 45234-02-4. A reported Ki of 1.3 mM for the PEPT1 membrane transporter is documented. It is provided as a high-purity reagent for biochemical and transporter studies.

  • Purity 98.0%.
  • Molecular weight 275.30 g/mol and formula C11H21N3O5.
  • Reported Ki 1.3 mM for PEPT1 transporter.
  • Available in multiple small-scale package sizes for laboratory use.
  • Intended for research use only; not for human consumption.
  • Suitable for peptide research, transporter assays, and biochemical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.