Learn More
Cayman Chemical Lysyllysyllysine (trifluoroacetate salt), Purity: ≥95%, 100 mg
SDP
Catalog No. 502909481
Encompass_Preferred
A tripeptide; has been conjugated to oleyl amine for the generation of cationic liposomes with anticancer activity
Catalog No. 50-290-9481 Supplier Cayman Chemical Supplier No. 36707100 MG
A tripeptide; has been conjugated to oleyl amine for the generation of cationic liposomes with anticancer activity