Learn More
Cayman Chemical Lysyllysyllysine (trifluoroacetate salt), Purity: ≥95%, 50 mg
SDP
Catalog No. 502913887
Encompass_Preferred
A tripeptide; has been conjugated to oleyl amine for the generation of cationic liposomes with anticancer activity
Catalog No. 50-291-3887 Supplier Cayman Chemical Supplier No. 3670750 MG
A tripeptide; has been conjugated to oleyl amine for the generation of cationic liposomes with anticancer activity