Learn More
Aviva Systems Biology Corp LYVE1 Antibody - N-terminal region (ARP46761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYVE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LYVE1. Alternate names: CRSBP-1, HAR, LYVE-1, XLKD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-59393