Learn More
AdipoGen Lyve1 pAb
SDP
Supplier:  AdipoGen AG25T0103C050
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse and rat LYVE-1. Source Rabbit. Applications ELISA, FACS, IHC, WB. Lyophilized. LYVE-1 has been identified as a major receptor for HA extracellular matrix glycosaminoglycan hyaluronan on the lymph vessel wall. Like CD44, the LYVE-1 molecule binds both soluble and immobilized HA. However, unlike CD44, the LYVE-1 molecule co-localizes with HA on the luminal face of the lymph vessel wall and is completely absent from blood vessels. Hence, LYVE-1 is the first lymph-specific HA receptor to be characterized and is a uniquely powerful marker for lymph vessels themselves.
Catalog No. 50-256-3170