Learn More
Aviva Systems Biology Corp LYZL4 Antibody - middle region (ARP62861_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62861_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYZL4. It was validated on Western Blot. . Alternate names: 1810009N24Rik, LYC4, MGC26768ApplicationWB. Species reactivity: Horse, Human, Rat
Catalog No. 50-287-789