Learn More
Sino Biological Recombinant Human MAP3K8 Protein (GST Tag)
Catalog No. 502574373
Encompass_Preferred
Recombinant human COT (30-397) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4373 Supplier Sino Biological Supplier No. M1615G100
Recombinant human COT (30-397) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.