Learn More
Aviva Systems Biology Corp LZTR1 Antibody - C-terminal region (ARP38863_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38863_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LZTR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LZTR1. Alternate names: LZTR-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53118