Learn More
Aviva Systems Biology Corp LZTR1 Antibody - N-terminal region (ARP38862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LZTR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LZTR1. Alternate names: LZTR-1, MGC21205, TCFL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56824