Learn More
Aviva Systems Biology Corp LZTS1 Antibody - C-terminal region (ARP39473_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39473_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LZTS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LZTS1. Alternate names: F37, FEZ1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57048