Learn More
Chemglass Life Sciences M-18 Threaded Adapter with Two 4mm Ports for Size 16 Pump Tubing
SDP
Catalog No. 501532113
Encompass
The double port adapter fits into the headplate and has two connections that are 4mm OD to fit precision tubing size 16.Inside the reactor, the 4mm tubes are 1.25" long.Stainless steel components are manufactured from 316L with an electropolished surface finish of 20Ra or better.
Catalog No. 50-153-2113 Supplier Chemglass Life Sciences Supplier No. CLS138057
The double port adapter fits into the headplate and has two connections that are 4mm OD to fit precision tubing size 16.Inside the reactor, the 4mm tubes are 1.25" long.Stainless steel components are manufactured from 316L with an electropolished surface finish of 20Ra or better.