Learn More
MSC 3M Versaflo Respiratory Hard Hat Assembly, Premium Visor, AS/NZS Head and Eye/Face Protection
Catalog No. 502835220
Encompass
The 3M Versaflo M-307 Respiratory Hard Hat Assembly provides integrated respiratory, eye, face, and head protection. It features a premium visor and faceseal, designed for challenging industrial environments. This lightweight helmet ensures comfort and compliance with safety standards.
  • Provides integrated respiratory, eye, face, and head protection.
  • Includes a premium visor and faceseal.
  • Lightweight and well-balanced design.
  • Fully-adjustable suspension ensures optimal fit.
  • Visor provides excellent peripheral and downward vision.
  • Coated polycarbonate visor resists chemicals and scratches.
  • Meets AS/NZS 1801 (head) and AS/NZS 1337.1 (eye/face) standards.
  • QRS breathing tubes for quick connection.
  • Removable parts simplify cleaning and maintenance.
  • Free from natural rubber latex.
Catalog No. 50-283-5220 Supplier MSC Supplier No. 41927153
The 3M Versaflo M-307 Respiratory Hard Hat Assembly provides integrated respiratory, eye, face, and head protection. It features a premium visor and faceseal, designed for challenging industrial environments. This lightweight helmet ensures comfort and compliance with safety standards.
  • Provides integrated respiratory, eye, face, and head protection.
  • Includes a premium visor and faceseal.
  • Lightweight and well-balanced design.
  • Fully-adjustable suspension ensures optimal fit.
  • Visor provides excellent peripheral and downward vision.
  • Coated polycarbonate visor resists chemicals and scratches.
  • Meets AS/NZS 1801 (head) and AS/NZS 1337.1 (eye/face) standards.
  • QRS breathing tubes for quick connection.
  • Removable parts simplify cleaning and maintenance.
  • Free from natural rubber latex.