Learn More
Apexbio Technology LLC m-3M3FBS 200933-14-8 50mg
Supplier: Apexbio Technology LLC B689350
m-3M3FBS (CAS 200933-14-8) is a potent activator of phospholipase C (PLC) widely used in studies of cell signaling pathways By stimulating PLC activity m-3M3FBS induces the hydrolysis of phosphoinositides leading to increased inositol phosphate production and elevated intracellular calcium concentrations Experimental data demonstrate that m-3M3FBS promotes superoxide generation in human neutrophils and enhances Ca mobilization supporting its utility in investigating oxidative burst and second messenger systems This small molecule is valuable for elucidating PLC-dependent mechanisms in signal transduction and related pathophysiological processes