Learn More
Medchemexpress LLC m-3M3FBS | 200933-14-8 | 99.99% | 343.36 | 100 MG
SDP
Catalog No. 5000400550
Encompass_Preferred
m-3M3FBS is a potent phospholipase C (PLC) activator. It stimulates superoxide generation in human neutrophils, upregulates intracellular calcium concentration, and stimulates inositol phosphate generation in various cell lines. It also induces monocytic leukemia cell apoptosis.
  • Stimulates superoxide generation in human neutrophils.
  • Upregulates intracellular calcium concentration.
  • Stimulates inositol phosphate generation in various cell lines.
  • Induces monocytic leukemia cell apoptosis.
Catalog No. 50-004-00550 Supplier Medchemexpress LLC Supplier No. HY19619100MG
m-3M3FBS is a potent phospholipase C (PLC) activator. It stimulates superoxide generation in human neutrophils, upregulates intracellular calcium concentration, and stimulates inositol phosphate generation in various cell lines. It also induces monocytic leukemia cell apoptosis.
  • Stimulates superoxide generation in human neutrophils.
  • Upregulates intracellular calcium concentration.
  • Stimulates inositol phosphate generation in various cell lines.
  • Induces monocytic leukemia cell apoptosis.