Learn More
Selleck Chemical LLC m-3M3FBS S0982-25MG
SDP
Catalog No. 5000042993
Encompass_Preferred
m-3M3FBS is a potent activator of phospholipase C (PLC) that stimulates a transient intracellular calcium concentration increase in neutrophils m-3M3FBS induces apoptosis in monocytic leukemia cells
Catalog No. 50-000-42993 Supplier Selleck Chemical LLC Supplier No. S098225MG
m-3M3FBS is a potent activator of phospholipase C (PLC) that stimulates a transient intracellular calcium concentration increase in neutrophils m-3M3FBS induces apoptosis in monocytic leukemia cells