Learn More
Medchemexpress LLC HY-19619 5mg Medchemexpress, m-3M3FBS CAS:200933-14-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196195MG
Encompass_Preferred
Medchemexpress, HY-19619 5mg m-3M3FBS CAS:200933-14-8 m-3M3FBS is a potent phospholipase C (PLC) activator. m-3M3FBS stimulates superoxide generation in human neutrophils, upregulates intracellular calcium concentration, and stimulates inositol phosphate generation in various cell lines. m-3M3FBS induces monocytic leukemia cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2150