Learn More
TARGETMOL CHEMICALS INC M-89 1MG
SDP
Catalog No. 502875921
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. M-89, a highly potent and specific inhibitor of the menin-mixed lineage leukemia (Menin-MLL) protein-protein interaction, exhibits a Kd of 1.4 nM for menin binding. This compound holds potential for treating MLL leukemia. Purity 99.58%
Catalog No. 50-287-5921 Supplier TARGETMOL CHEMICALS INC Supplier No. T119251MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. M-89, a highly potent and specific inhibitor of the menin-mixed lineage leukemia (Menin-MLL) protein-protein interaction, exhibits a Kd of 1.4 nM for menin binding. This compound holds potential for treating MLL leukemia. Purity 99.58%