Learn More
Medchemexpress LLC M-89 | 2363165-42-6 | 99.1% | 50 MG
SDP
Catalog No. 5000340417
Encompass_Preferred
M-89 is a highly potent and specific menin inhibitor that targets the menin-mixed lineage leukemia (Menin-MLL) protein-protein interaction. It has potential for studying MLL leukemia by inhibiting cell growth in leukemia cell lines and inducing apoptosis. It is for research use only.
  • Highly potent and specific menin inhibitor
  • Inhibits menin-MLL protein-protein interaction
  • Potential for studying MLL leukemia
  • Inhibits cell growth in leukemia cell lines carrying MLL fusion
  • Induces time- and dose-dependent apoptosis in MV4;11 cells
  • Stabilizes cellular menin protein in MV4;11 and MOLM-13 cells
Catalog No. 50-003-40417 Supplier Medchemexpress LLC Supplier No. HY12834750MG
M-89 is a highly potent and specific menin inhibitor that targets the menin-mixed lineage leukemia (Menin-MLL) protein-protein interaction. It has potential for studying MLL leukemia by inhibiting cell growth in leukemia cell lines and inducing apoptosis. It is for research use only.
  • Highly potent and specific menin inhibitor
  • Inhibits menin-MLL protein-protein interaction
  • Potential for studying MLL leukemia
  • Inhibits cell growth in leukemia cell lines carrying MLL fusion
  • Induces time- and dose-dependent apoptosis in MV4;11 cells
  • Stabilizes cellular menin protein in MV4;11 and MOLM-13 cells