Learn More
Medchemexpress LLC M-89 | 2363165-42-6 | 99.1% | 657.87 g/mol | C37H47N5O4S | 5 MG
SDP
Catalog No. 5000264343
Encompass_Preferred
M-89 is a potent, selective menin-MLL interaction inhibitor supplied as a research compound for preclinical biochemical and cellular studies. It is used to disrupt the menin-MLL protein-protein interaction in epigenetic and leukemia research.
  • Potent menin-MLL protein-protein interaction inhibitor (Kd 1.4 nM).
  • High purity (99.1%) suitable for biochemical assays.
  • Soluble in DMSO at approximately 7.35 mg/mL with ultrasonic assistance.
  • Available in small research quantities for exploratory studies.
  • Stable when stored as powder at low temperatures for long-term use.
Catalog No. 50-002-64343 Supplier Medchemexpress LLC Supplier No. HY1283475MG
M-89 is a potent, selective menin-MLL interaction inhibitor supplied as a research compound for preclinical biochemical and cellular studies. It is used to disrupt the menin-MLL protein-protein interaction in epigenetic and leukemia research.
  • Potent menin-MLL protein-protein interaction inhibitor (Kd 1.4 nM).
  • High purity (99.1%) suitable for biochemical assays.
  • Soluble in DMSO at approximately 7.35 mg/mL with ultrasonic assistance.
  • Available in small research quantities for exploratory studies.
  • Stable when stored as powder at low temperatures for long-term use.