Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich m-Anisaldehyde

Catalog No. 12965825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-965-825G 25 g
12-965-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-965-825G Supplier Sigma Aldrich Supplier No. 12965825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • M-anisaldehyde has significant antifungal activity against candida sp., including the azole-resistant strains.m-anisaldehyde was used as an eluent for mono-13c isotopomers of vanillin in normal phase silica gel chromatography
  • It was also used as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (nnk) metabolism

Specifications

CAS 591-31-1
Density 1.117 g/mL (at 20°C (literature))
Boiling Point 143°C (50 mmHg)
Molecular Formula C8H8O2
Linear Formula CH3OC6H4CHO
Refractive Index n20/D 1.553 (literature)
MDL Number MFCD00003361
Quantity 25 g
Synonym 3-Methoxybenzaldehyde
Molecular Weight (g/mol) 136.15
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.