Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-Anisaldehyde (3-methoxybenzaldehyde) | 591-31-1 | 136.15 | 50 G
SDP

Supplier:  Medchemexpress LLC HYW00734650G

Encompass_Preferred

m-Anisaldehyde (3-Methoxybenzaldehyde) is an endogenous metabolite appearing as a colorless to light yellow liquid with a density of 1.117 g/cm³. Its chemical formula is C8H8O2, and it is classified under Ketones, Aldehydes, and Acids, primarily sourced from plants.

  • Endogenous metabolite associated with thyroid autoimmunity during early pregnancy
  • Colorless to light yellow liquid
  • Density: 1.117 g/cm³
  • Chemical formula: C8H8O2
  • Classified under ketones, aldehydes, and acids
  • Primarily sourced from plants
  • Recommended storage at 4°C under nitrogen
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-60400


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.