Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-Anisaldehyde | 591-31-1 | 136.15 | 100 G
SDP

Catalog No. 5000460259
Encompass_Preferred

m-Anisaldehyde (3-Methoxybenzaldehyde) is an endogenous metabolite with a molecular weight of 136.15 and a purity of 99.65%. It is a colorless to light yellow liquid associated with thyroid autoimmunity during early pregnancy. This product is intended for research use only.

  • Endogenous metabolite
  • Associated with thyroid autoimmunity during early pregnancy
  • Colorless to light yellow liquid appearance
  • Purity of 99.65%
  • Store at 4°C under nitrogen
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-60259 Supplier Medchemexpress LLC Supplier No. HYW007346100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-Anisaldehyde (3-Methoxybenzaldehyde) is an endogenous metabolite with a molecular weight of 136.15 and a purity of 99.65%. It is a colorless to light yellow liquid associated with thyroid autoimmunity during early pregnancy. This product is intended for research use only.

  • Endogenous metabolite
  • Associated with thyroid autoimmunity during early pregnancy
  • Colorless to light yellow liquid appearance
  • Purity of 99.65%
  • Store at 4°C under nitrogen
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.