Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure Mouse Anti-Human IgG, Fcγ fragment specific, 1ml
SDP
Catalog No. NC9122619
Encompass_Preferred
Peroxidase-AffiniPure Mouse Anti-Human IgG, Fc Fragment Specific (min X Bov,Hrs,Ms Sr Prot)
Catalog No. NC9122619 Supplier Jackson Immuno Research Labs Supplier No. 209035098
Peroxidase-AffiniPure Mouse Anti-Human IgG, Fc Fragment Specific (min X Bov,Hrs,Ms Sr Prot)