Learn More
TWD Scientific LLC Medium Antistatic Polypropylene Weighing Funnel. Pack of 50.
SDP
Catalog No. 501173379 Shop All TWD Scientific LLC Products
Encompass_Preferred
Medium disposable antistatic polypropylene weighing funnel. Fits volumetric flask volumes (ml) of 100, 200, 250. Funnels have a maximum capacity of 10mL solid/7mL liquid. 50/Pack
Catalog No. 50-117-3379 Supplier TWD Scientific LLC Supplier No. ASWF2MPK
Medium disposable antistatic polypropylene weighing funnel. Fits volumetric flask volumes (ml) of 100, 200, 250. Funnels have a maximum capacity of 10mL solid/7mL liquid. 50/Pack