Learn More
Electron Microscopy Sciences M-Bond 43B Kit
SDP
Catalog No. 509053762
Encompass_Preferred
The M-Bond adhesives are high-performance epoxy resins, which are formulated especially for bonding strain gages and special-purpose sensors. When properly cured, these adhesives are useful in temperatures ranging from -452F to +350 F (-269C to+ 175C) with M-Bond 43-B, and to +700F (+370C) for short periods with M-bond 600 and 610. In common with other organic materials, oxidation and sublimation effects at elevated temperatures limit life. Shelf Life: 9 months.
Curing time: 2 hours at +375F (+190C)
Package: 4 x 25 ml
Catalog No. 50-905-3762 Supplier Electron Microscopy Sciences Supplier No. 5041010
The M-Bond adhesives are high-performance epoxy resins, which are formulated especially for bonding strain gages and special-purpose sensors. When properly cured, these adhesives are useful in temperatures ranging from -452F to +350 F (-269C to+ 175C) with M-Bond 43-B, and to +700F (+370C) for short periods with M-bond 600 and 610. In common with other organic materials, oxidation and sublimation effects at elevated temperatures limit life. Shelf Life: 9 months.
Curing time: 2 hours at +375F (+190C)
Package: 4 x 25 ml