Learn More
Bioworld M-Broth, 500 g
Catalog No. 50254458
Encompass_Preferred
M-BrothUsed in detecting Salmonella sp. in foods and feeds by the accelerated enrichment serology (ES) procedurepH: 7.0Dehydrated Appearance: Beige, homogeneous powderPrepared Appearance: light amber broth; may have slight precipitateSoluble in boiling waterComposition:Pancreatic Digest of Casein Yeast Extract D-Mannose Sodium Citrate Sodium Chloride Dipotassium Phosphate Manganese Chloride Magnesium Sulfate Ferrous Sulfate Tween 80
Catalog No. 50-254-458 Supplier Bioworld Supplier No. 306230011
M-BrothUsed in detecting Salmonella sp. in foods and feeds by the accelerated enrichment serology (ES) procedurepH: 7.0Dehydrated Appearance: Beige, homogeneous powderPrepared Appearance: light amber broth; may have slight precipitateSoluble in boiling waterComposition:Pancreatic Digest of Casein Yeast Extract D-Mannose Sodium Citrate Sodium Chloride Dipotassium Phosphate Manganese Chloride Magnesium Sulfate Ferrous Sulfate Tween 80