Learn More
NEOGEN m-Broth, 500 g
SDP
Catalog No. 502015513
Encompass_Preferred
M Broth is used for the cultivation of Salmonella spp in a laboratory setting. M-Broth is not intended for use in the diagnosis of disease or other conditions in humans. M-Broth is prepared according to the formula of Sperber and Diebel, and contains the nutrients necessary for good growth and flagella development of Salmonella. This medium is used for cultivating Salmonella in foods and feeds by the accelerated enrichment serology (ES) procedure. M-Broth conforms to standard procedures recommended in food testing and monoclonal and polyclonal enzyme immunoassays (EIA).For more information visit https://www.neogen.com/solutions/Microbiology/m-broth
Catalog No. 50-201-5513 Supplier NEOGEN Supplier No. 700003332
M Broth is used for the cultivation of Salmonella spp in a laboratory setting. M-Broth is not intended for use in the diagnosis of disease or other conditions in humans. M-Broth is prepared according to the formula of Sperber and Diebel, and contains the nutrients necessary for good growth and flagella development of Salmonella. This medium is used for cultivating Salmonella in foods and feeds by the accelerated enrichment serology (ES) procedure. M-Broth conforms to standard procedures recommended in food testing and monoclonal and polyclonal enzyme immunoassays (EIA).For more information visit https://www.neogen.com/solutions/Microbiology/m-broth