Learn More
Lifeline Cell Technology M BTCELL HU LIFELINE KT
SDP
Catalog No. 50311899
Encompass_Preferred
Includes BronchiaLife Basal Medium 485 mL, BronchiaLife B/T LifeFactors Kit. Optimized for the serum-free culture of human airway epithelial cells. Supports the growth of these cells over a period of at least 15 population doublings.
Catalog No. 50-311-899 Supplier Lifeline Cell Technology Supplier No. LL0023
Includes BronchiaLife Basal Medium 485 mL, BronchiaLife B/T LifeFactors Kit. Optimized for the serum-free culture of human airway epithelial cells. Supports the growth of these cells over a period of at least 15 population doublings.