Learn More
Bellco Glass, Inc. MicroCarrier Flask Only,500mL,100mm Nk W/Dimple Bottom
SDP
Catalog No. 501944996
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. SIZE: 500ml (working volume). Dimensions: 110mm OD x 170mm overall height. Center neck: 100mm. Two 45mm angled sidearms; (ANGLED 60°). Grad. Range: 100-500ml, every 50ml. Supply with (2) 45mm PP blue screw caps & pouring rings.
Catalog No. 50-194-4996 Supplier Bellco Glass, Inc. Supplier No. 196520500
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. SIZE: 500ml (working volume). Dimensions: 110mm OD x 170mm overall height. Center neck: 100mm. Two 45mm angled sidearms; (ANGLED 60°). Grad. Range: 100-500ml, every 50ml. Supply with (2) 45mm PP blue screw caps & pouring rings.