Learn More
Bellco Glass, Inc. MicroCarrier Impeller Assembly , 1-3L
SDP
Catalog No. 501945011
Encompass_Preferred
Micro-Carrier ImpellerAssemblyPromote optimal growth of suspension andmicro-carrier cultures. Impeller height adjuststhrough compression fitting in cap. Indentedbottom up to 1L prevents accumulation ofmicro-carrier beads under impeller. Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 1-3l Paddle dimensions: 1 3/4" x 4 1/4" Consist of: Magnet holder: a523-207 Silicone plug: a523-200
Catalog No. 50-194-5011 Supplier Bellco Glass, Inc. Supplier No. 196531000
Micro-Carrier ImpellerAssemblyPromote optimal growth of suspension andmicro-carrier cultures. Impeller height adjuststhrough compression fitting in cap. Indentedbottom up to 1L prevents accumulation ofmicro-carrier beads under impeller. Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 1-3l Paddle dimensions: 1 3/4" x 4 1/4" Consist of: Magnet holder: a523-207 Silicone plug: a523-200