Learn More
Bellco Glass, Inc. MicroCarrier Impeller Assembly , 100-250mL
SDP
Catalog No. 501945009
Encompass_Preferred
Micro-Carrier ImpellerAssemblyPromote optimal growth of suspension andmicro-carrier cultures. Impeller height adjuststhrough compression fitting in cap. Indentedbottom up to 1L prevents accumulation ofmicro-carrier beads under impeller.. Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 100-250ml Paddle dimensions: 1" x 2"
Catalog No. 50-194-5009 Supplier Bellco Glass, Inc. Supplier No. 196530100
Micro-Carrier ImpellerAssemblyPromote optimal growth of suspension andmicro-carrier cultures. Impeller height adjuststhrough compression fitting in cap. Indentedbottom up to 1L prevents accumulation ofmicro-carrier beads under impeller.. Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 100-250ml Paddle dimensions: 1" x 2"