Learn More
Bellco Glass, Inc. MicroCarrier Impeller Assembly , 15/36L
SDP
Catalog No. 501945013
Encompass_Preferred
Micro-Carrier ImpellerAssemblyPromote optimal growth of suspension andmicro-carrier cultures. Impeller height adjuststhrough compression fitting in cap. Indentedbottom up to 1L prevents accumulation ofmicro-carrier beads under impeller. Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 15-36l Paddle dimensions: 3"x 10"
Catalog No. 50-194-5013 Supplier Bellco Glass, Inc. Supplier No. 196531500
Micro-Carrier ImpellerAssemblyPromote optimal growth of suspension andmicro-carrier cultures. Impeller height adjuststhrough compression fitting in cap. Indentedbottom up to 1L prevents accumulation ofmicro-carrier beads under impeller. Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 15-36l Paddle dimensions: 3"x 10"