Learn More
Bellco Glass, Inc. MicroCarrier Impeller Assembly , 6-8L
SDP
Catalog No. 501945021
Encompass_Preferred
Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 6-8l Paddle dimensions: 3"x 7 3/4"
Catalog No. 50-194-5021 Supplier Bellco Glass, Inc. Supplier No. 196538000
Impeller assembly for micro-carrier spinner flasks. Consist of magnet holder, silicone plug, magnet, and ptfe paddle. MADE IN THE USA. Impeller size: 6-8l Paddle dimensions: 3"x 7 3/4"