Learn More
Bio IVT M C-MONK CRY HEP 5 M CLLSV
Catalog No. 502310855
Encompass
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Cynomolgus monkey, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial
This item is not returnable. View return policy
Catalog No. 50-231-0855 Supplier Bio IVT Supplier No. M00305
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Cynomolgus monkey, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial